(5R)-6,6'-Bis(bis(3,5-dimethylphenyl)phosphaneyl)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine ≥97%

Supplier: Ambeed
Warning

A1475931-1G
77597-490EA 780.03 USD
77597-490
(5R)-6,6'-Bis(bis(3,5-dimethylphenyl)phosphaneyl)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine ≥97%
(5R)-6,6'-Bis(bis(3,5-dimethylphenyl)phosphaneyl)-2,2',3,3'-tetrahydro-5,5'-bibenzo[b][1,4]dioxine

Precaución: for Research Use Only

Formula: C₄₈H₄₈O₄P₂
MW: 750.8300 g/mol
MDL Number: MFCD34598587
CAS Number: 1306747-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR