Ethyl pent-4-ynoate ≥97%

Supplier: Ambeed
Danger

Synonyms: 4-Pentynoic Acid Ethyl Ester

A961576-5G A961576-1G
77573-902EA 28 USD
77573-902 77595-554
Ethyl pent-4-ynoate ≥97%
Ethyl pent-4-ynoate

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.1531 g/mol
MDL Number: MFCD18976214
CAS Number: 63093-41-4
UN: 3272
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR