5'-Fluoro-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

A159065-100G A159065-10G A159065-1G A159065-25G A159065-500G
103843-438EA 6 USD
103843-438 77235-782 77235-784 77235-786 77319-312 77592-934
5'-Fluoro-2'-hydroxyacetophenone 98%
5'-Fluoro-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 182.2 °C (760 mm Hg)
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00011668
CAS Number: 394-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR