4-Chlorothieno[2,3-b]pyridine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A1481606-1G
77592-782EA 382.33 USD
77592-782
4-Chlorothieno[2,3-b]pyridine-2-carboxylic acid ≥95%
4-Chlorothieno[2,3-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂S
MW: 213.6409 g/mol
MDL Number: MFCD11847968
CAS Number: 1379335-94-0
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR