Thermo Scientific™ Capit-All™ Flex Automated Decapper
Supplier: Thermo Fisher Scientific
4120-FLEX
77592-016EA 75610.13 USD
77592-016
Thermo Scientific™ Capit-All™ Flex Automated Decapper
Crimpers, Cappers and Decappers
A full-service, automated capping and decapping solution for Thermo Scientific™ Matrix™ and Nunc™ 48 and 96 tube racks.
  • Flexibility: Designed for use as a standalone benchtop unit or in an integrated system
  • Compatible with Matrix and Nunc tubes in 48- and 96-tube formats, can switch between tube formats without adapters, supporting both current and changing application requirements
  • Efficiency: Completes a capping cycle in less than 60 seconds, can handle both full and partial racks of tubes, offering flexibility for varying workflows
  • Angled, 7" touchscreen
More Product Information