CE MicroIntensive Duo® Plus L Mop
Supplier: Vileda Professional - FHP
172281
77591-710CS 297.07 USD
77591-710
CE MicroIntensive Duo® Plus L Mop
Mops
The CE MicroIntensive Duo® Plus product family is a single-use mop specifically for effective IPA rinsing and the removal of disinfectant residue on glass and stainless steel.
  • High surface coverage with double-sided design
  • Effective residue removal on glass and stainless steel
  • Ideal for damp mopping and pre-preparation cleaning methods
  • Streak free cleaning
  • Extremely low particle shedding - Up to ISO 4 (GMP A/B); IPA compatible
  • Saves time, money, and chemical use
  • Laundered
High performance dimpled polyester microfiber for excellent cleaning results.