LabExact® PES Syringe Filters with Glass Fiber Prefilter
Supplier: I.W. TREMONT CO., INC.
LEES10206 LEES10207 LEES10208 LEES10209
77591-026PK 244.51 USD
77591-026 77591-028 77591-030 77591-032
LabExact® PES Syringe Filters with Glass Fiber Prefilter
Filters Syringe Filters
LabExact® disposable non sterile PES/Polyethersulfone hydrophilic filtration media syringe filters have high filtration flow rates and an inherently low affinity for proteins and 1 µm glass microfiber prefilter to raises absorption capacity and loading.
  • Low extractables and ideal for applications requiring maximum sample recovery
  • Use for introduction of additives into tissue culture vessel and food, beverage and ultra pure water final filtration
  • Use for final filtration of bacteriophage and infusion solutions and tissue culture, proteins, buffer and
More Product Information