LabExact® PTFE Syringe Filters, Non Sterile
Supplier: I.W. TREMONT CO., INC.
LEES10036 LEES10037 LEES10017 LEES10018 LEES10019 LEES10020 LEES10034 LEES10035 LEES10320 LEES10321 LEES10322 LEES10323 LEES10603 LEES10604 LEES10605 LEES10606
77590-966PK 256.09 USD
77590-966 77590-968 77590-954 77590-956 77590-958 77590-960 77590-962 77590-964 77591-066 77591-068 77591-070 77591-072 77591-102 77591-104 77591-106 77591-108
LabExact® PTFE Syringe Filters, Non Sterile
Filters Syringe Filters
LabExact® disposable non sterile PTFE hydrophobic filtration media syringe filters are inherently hydrophobic; both hydrophobic and hydrophilic syringe filters are ideal for the filtration of nonpolar liquids and gases and are very resistant to solvents as well as acids or bases.
  • Excellent chemical compatibility for filtering harsh chemicals that destroy other membrane materials
  • Use for filtering and de-gassing chromatography solvents or filtering viscous organic-based HPLC samples
  • Ideal for aerosol sampling and can be used as a moisture barrier for
More Product Information