Cover for Rack 'Series 20'
Supplier: SARSTEDT INC
93.1102.001
77589-608CS 434.46 USD
77589-608
Cover for Rack 'Series 20'
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Lid ensures secure sample storage
  • Can be autoclaved at 121 °C
  • For storage down to −86 °C
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.