Serological Pipettes, Graduated, Individually Wrapped
Supplier: SARSTEDT INC
86.1254.001 86.1256.001 86.1251.001 86.1252.001 86.1253.001 86.1685.001
77589-564CS 287.49 USD
77589-564 77589-568 77589-546 77589-550 77589-558 77589-584
Serological Pipettes, Graduated, Individually Wrapped
Pipettes Serological Pipettes
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
  • Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Easy volume identification using the international color code on each pipette and blister packaging
  • Optimized mouthpieces for universal and secure fit in common pipetting aids
  • Larger pipetting volume due to negative scaling
  • Variable
More Product Information