(Safety-) Multifly® Needles
Supplier: SARSTEDT INC
85.1640.235 85.1642.205 85.1642.235 85.1638 85.1638.005 85.1638.035 85.1638.200 85.1638.205 85.1638.235 85.1638.935 85.1640.005 85.1640.200 85.1640.205
77589-482CS 808.74 USD
77589-482 77589-484 77589-486 77589-488 77589-458 77589-460 77589-462 77589-464 77589-466 77589-468 77589-470 77589-472 77589-474 77589-476 77589-478 77589-480
(Safety-) Multifly® Needles
Syringe Needles
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • Safety butterfly needles with integrated holder and non-safety needle options
  • Ready to use immediately - no complicated assembly of needle and holder required
  • DEHP-free - for short-term infusion
  • Flat puncturing angle ensures safe puncture
More Product Information