Screw Cap for Urine Collection Cups
Supplier: SARSTEDT INC
76.564
77589-086CS 123.46 USD
77589-086
Screw Cap for Urine Collection Cups
Closures Test/Sample Tube Closures
To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: HDPE
  • Compatible with screw cap for 100 ml urine cup (77589-064)
  • Odor-tight screw cap
  • For secure transport and storage of urine samples
Small volume urine containers for consistent and reliable samples.