Urine Cups with Screw Cap
Supplier: SARSTEDT INC
75.563 75.562.005 75.562.011 75.562.105 75.562.300
77589-064CS 185.77 USD
77589-064 77589-052 77589-054 77589-056 77589-058
Urine Cups with Screw Cap
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Small volume urine containers for consistent and reliable samples.
  • Material: PP
  • Transparent material or with light protection property (brown)
  • Odor-tight screw cap
  • With graduations and labelling area
  • Suitable for transport and storage of urine samples
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, SARSTEDT offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.