Screw Cap Microtubes
Supplier: SARSTEDT INC
72.733.010 72.733.201 72.785 72.785.005 72.694.396 72.694.300 72.694.305 72.694.306 72.694.307 72.694.310 72.694.321 72.694.322 72.694.323 72.694.325 72.694.328 72.730.026 72.730.027 72.730.028 72.730.100 72.730.105 72.730.106 72.730.107 72.730.300 72.730.303 72.703 72.703.004 72.693 72.693.005 72.693.100 72.693.105 72.694.100 72.694.105 72.694.106 72.694.107 72.607 72.607.772 72.608 72.609.300 72.609.303 72.609.311 72.609.601 72.664 72.730.024 72.730.025 72.730.304 72.730.305 72.730.306 72.730.307 72.730.308 72.730.309 72.730.311 72.733.001 72.664.711 72.687 72.687.772 72.692 72.692.005 72.692.100 72.692.105 72.692.995
77588-982CS 836 USD
77588-982 77588-984 77588-998 77589-000 77588-822 77588-802 77588-804 77588-806 77588-808 77588-810 77588-812 77588-814 77588-816 77588-818 77588-820 77588-942 77588-944 77588-946 77588-948 77588-950 77588-952 77588-954 77588-958 77588-960 77588-878 77588-880 77588-782 77588-784 77588-786 77588-788 77588-792 77588-794 77588-796 77588-798 77588-686 77588-688 77588-690 77588-692 77588-694 77588-696 77588-698 77588-700 77588-938 77588-940 77588-962 77588-964 77588-966 77588-968 77588-970 77588-972 77588-974 77588-980 77588-702 77588-704 77588-706 77588-766 77588-768 77588-770 77588-772 77588-780
Screw Cap Microtubes
Tubes Microtubes
SARSTEDT screw cap microtubes provide an exceptionally secure seal with the integrated O-ring. Ideal for centrifugation, transport and storage of high-quality oligonucleotides, enzymes and buffers.
  • Material: PP
  • Robust polypropylene material for safety and versatility
  • 3 different volumes meet filling requirements from a few microliters to 2 milliliters
  • Leak-resistant according to ADR and IATA- tested at a vacuum of 0.95 bar
  • Centrifugation up to 20000×g
  • Can be autoclaved to 121 °C and frozen to –80 °C
  • O-ring cap increases cap security (IATA tested)
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable