Screw Cap Tubes with Skirted Conical Base, PS
Supplier: SARSTEDT INC
60.9921.821 60.9921.829 60.9921.830
77587-832CS 290.36 USD
77587-832 77587-834 77587-836
Screw Cap Tubes with Skirted Conical Base, PS
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.
  • Material: PS
  • Graduations: No
  • Base shape: Skirted conical base
  • Corresponds to the requirement: ADR
  • Tube with skirted base ensures that it stands securely
  • Polystyrene (PS) tubes for maximum transparency
  • Especially secure cap
This broad product portfolio in the field of reagent and centrifuge tubesMore Product Information