Prepared Micro Sample Tubes with Push Caps
Supplier: SARSTEDT INC
41.1500.005 41.1501.105 41.1503.105 41.1504.105 41.1506.100 41.1506.102
77587-468CS 819.27 USD
77587-468 77587-470 77587-472 77587-474 77587-476 77587-478
Prepared Micro Sample Tubes with Push Caps
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
SARSTEDT prepared micro tubes are available with push cap closure or screw cap with integral O-ring.
  • Ø: 10.8 mm
  • Length without/with cap: 40/44 mm
  • Base: Skirted conical base
  • Ideal collection vessel for small volumes of blood
  • Attached push cap suitable for one-handed collection technique
  • Transparent label with graduations ideal for checking fill level; available with paper label (*.015) on request
These micro volume tubes are prepared with standard additives for versatile low-volume requirements.