Microvette® with Capillary
Supplier: SARSTEDT INC
20.1278.100 20.1280.100 20.1282.100 20.1288.100 20.1290.100 20.1291 20.1292.100
MSPP-20128810PK 63.93 USD
MSPP-20128810 102971-718 103093-113 77587-418 77587-420 77587-422 77587-424 77587-426 77587-428 77587-430
Microvette® with Capillary
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Capillary blood collection facilitates diagnostics with a small volume of blood.
  • Closure: Cap
  • Outer container with skirted base and graduations
  • End-to-end capillary pre-assembled
  • Smooth cap - special lid design reduces the aerosol effect
  • Conical inner tube: After centrifugation, higher supernatant for easier collection of plasma - cylindrical inner tube: Optimal tube shape ensures the sample is well mixed
  • Two collection techniques: Capillary or collection rim
The capillary blood collection systems are among the most versatile on the market. Microvette®More Product Information