End-to-end Capillaries
Supplier: SARSTEDT INC
19.447.001
77587-414CS 6514.75 USD
77587-414
End-to-end Capillaries
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Capillary blood collection facilitates diagnostics with a small volume of blood. SARSTEDT offers innovative solutions for all patient groups.
  • Material: Glass
  • Volumes: 10 and 20 µl
  • Preparations: Ammonium heparin, EDTA
Simple and safe capillary blood collection.