BradyJet™ J7300 Color Label Printer WiFi with Safety and Facility ID Software
Supplier: Brady Worldwide
176877 176878
77581-068EA 2392.21 USD
77581-068 77580-896
BradyJet™ J7300 Color Label Printer WiFi with Safety and Facility ID Software
Printers Label Printers
Unleash true color with the BradyJet™ J7300 Color label printer. Create vibrant, industrial-grade inkjet for safety labels, signs, and tags in full color. Non-WiFi option available.
  • Full color printing capability
  • PC connected
  • Print resolution: 4800 dpi
  • 2 year warranty
  • Maximum label width: 4.25"
  • Maximum labels per day: 5000
Ideal for industrial safety labeling those changes frequently, from Lean/5S, pipe marking and lockout tagout to OSHA/ANSI signage and custom labels. LabelSense™ technology enables printer, materials, and inks to talk to each otherMore Product Information