X-ray and Metal Detectable Powder Funnels
Supplier: SAMPLING SYSTEMS LTD
M1051B-200
77578-224EA 42.85 USD
77578-224
X-ray and Metal Detectable Powder Funnels
Funnels Powder Funnels
X-ray and metal detectable powder funnels are ideal for both laboratory and production work. The bright blue colour makes the bottles easy to spot if they are left in the product.
  • Robust PP construction
  • Non sterile
  • PP gives excellent chemical resistance to many products
  • Full batch traceability
To ensure the cleanliness of the funnels, they are manufactured and packed in a cleanroom. The robust nature of these funnels means that they can give many years of service or for more demanding applications. They can be used as a single use disposable productMore Product Information