Drums, Stainless Steel, with Toggle Clamps
Supplier: SAMPLING SYSTEMS LTD
A626-1 A626-2 A626-3
77578-076EA 1155.28 USD
77578-076 77578-078 77578-080
Drums, Stainless Steel, with Toggle Clamps
Barrels, Kegs, Drums
This popular container range is available in a range of sizes from 1 to 15 L. They are manufactured from 316L stainless steel and finished to a high quality finish. Ideal for work in a range of different environments include small scale production, laboratories and general day to day use.
  • They are ideal for working with both powders and liquids
  • The containers come with a drop-on lid
  • The lid is secured with toggle clamps and an O-ring seal
  • Simply pull down the toggle clamps to secure the lid
  • Storage conditions: Dry and ambient temperature