Chemical Spoon and Spatulas
Supplier: SAMPLING SYSTEMS LTD
A625-150 A625-280 A625-320
77578-070EA 6.38 USD
77578-070 77578-072 77578-074
Chemical Spoon and Spatulas
Spoons
Versatile spoon and spatula for sampling powder and liquid products.
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Can be used as both a spoon and a spatula
  • Perfect for measuring, sampling and dosing powders and liquids
This versatile product has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula allowing greater versatility in the sampling of both powder and liquid products. There are no sharp edges or crevices and are ideal for use in food, pharma and chemical sectors.