Mini-Pots, with Lid, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A426-500 A426-100 A426-1000 A426-25 A426-250 A426-50 A426-7
77577-946EA 429.3 USD
77577-946 77577-932 77577-934 77577-940 77577-942 77577-944 77577-950
Mini-Pots, with Lid, Stainless Steel
Trays/Dishes/Pots Pots
Made of 316L stainless steel.
  • Crevice free interior
  • Screw on lid
  • Very good chemical and temperature resistance
Multi purpose small volume containers with satin finish.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable