Pallet Knife, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A330-250 A330-130
77577-866EA 37.49 USD
77577-866 77577-864
Pallet Knife, Stainless Steel
Knives
These all stainless steel Pallet Knives are ideal for use in various industries such as the chemical and food industries. Each Pallet Knife is made from stainless steel with a 420 grade stainless steel blade.
  • Made from a high quality stainless steel
  • Wide range of sizes available from stock
We use 420 grade stainless steel as it is flexible and has excellent corrosion resistance. Other materials like 304 and 316 stainless steel do not have the flexibility.
The all stainless steel design means the pallet knives are easy to use and clean. The polishedMore Product Information