ULTRA Micro Spatulas, USP Class VI, Double-Bagged
Supplier: SAMPLING SYSTEMS LTD
9015H-170S 9015HN-170S 9015HV-170S
77577-564EA 158.01 USD
77577-564 77577-566 77577-568
ULTRA Micro Spatulas, USP Class VI, Double-Bagged
Spatulas
The double bagged micro spatula are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in class 100,000 cleanroom, packed in Class 10,000 cleanroom
Simply unwrap and use the micro spatula. The narrow design means they can be used with most vials and test tubes.
This range is ideal for sampling and dispensing small amounts.
Certificaciones: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661.
Envase: Supplied in a lined box.