Fluid Point Sampler
Supplier: SAMPLING SYSTEMS LTD
5551S-1000 5551S-1500 5551S-600
77576-572EA 739.45 USD
77576-572 77576-574 77576-576
Fluid Point Sampler
Samplers Fluid Samplers
The fluid point sampler is designed for taking rapid samples of free-flowing liquids.
  • Ideal for sampling free-flowing liquids from most containers
  • Great for sampling pharmaceuticals, cosmetics, and foodstuffs
  • Easily cleaned
  • Available from stock
  • Fast delivery
The fluid point sampler is great for taking samples of free-flowing liquids from a range of containers, including drums. It is made from chemically resistant 316L stainless steel to provide excellent corrosion resistance.