Cleanroom Arc Flash Booties
Supplier: Sellstrom
CR-BOOTIE-LARGE CR-BOOTIE-MED CR-BOOTIE-SMALL
77575-486EA 911.64 USD
77575-486 77575-496 77575-518
Cleanroom Arc Flash Booties
Workwear
Clean room arc rated booties are designed to meet specific cleanliness requirements for use in clean room environments.
  • Booties are particle-free
  • Made from non-shedding materials (flame-resistant Nomex fabric) to prevent contamination of the environment
  • Provide protection against arc flash hazards and electrostatic discharge
  • Generous fit for maximum comfort while performing electrical work tasks
  • Dimension: 10×8×6"
These garments may be worn by workers in industries such as semiconductor manufacturing, pharmaceuticals, and electronics assembly, amongMore Product Information