Vacuum Pumps, V-180
Supplier: Buchi
11V180000 11V180001 11V180010 11V180011 11V180100 11V180101 11V180110 11V180111 11V180200 11V180201 11V180210 11V180211
77569-912EA 3289.44 USD
77569-912 77569-914 77569-916 77569-918 77569-920 77569-922 77569-924 77569-926 77569-928 77569-930 77569-932 77569-934
Vacuum Pumps, V-180
Pumps Vacuum Pumps
The vacuum pump V-180 is a chemically resistant PTFE diaphragm vacuum pump with space saving design. It covers essential needs in vacuum generation.
  • Effective vacuum source for limited budgets
  • Small footprint saves valuable laboratory space
  • Light weight and compact design with removable carrying handle
  • High up time due to low maintenance
  • Accurate vacuum regulation thanks to optional interface
  • Robust and stable design and Integrated ECO2 mode
  • On/off-regulated by the Interface I-180, optimally suited to work with Rotavapor® R-80
Speed-controlled vacuumMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable