6-Amino-1,3-dimethyl-5-nitrosopyrimidine-2,4(1H,3H)-dione ≥98% (water<30%)

Supplier: Ambeed
Warning

A1996953-25G
77567-874EA 11.33 USD
77567-874
6-Amino-1,3-dimethyl-5-nitrosopyrimidine-2,4(1H,3H)-dione ≥98% (water<30%)
6-Amino-1,3-dimethyl-5-nitrosopyrimidine-2,4(1H,3H)-dione

Precaución: For research use only.

Formula: C₆H₈N₄O₃
MW: 184.153 g/mol
MDL Number: MFCD00023799
CAS Number: 6632-68-4
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR