4-(1-Methylcyclopropyl)aniline hydrochloride 98%

Supplier: Ambeed
Warning

A1421498-250MG
77566-862EA 371.83 USD
77566-862
4-(1-Methylcyclopropyl)aniline hydrochloride 98%
4-(1-Methylcyclopropyl)aniline hydrochloride

Precaución: For research use only.

Formula: C₁₀H₁₄ClN
MW: 183.678 g/mol
MDL Number: MFCD31616179
CAS Number: 2172473-99-1
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR