1-Bromo-4-(trimethylsilyl)but-3-yn-2-one 95%

Supplier: Ambeed
Danger

A1080385-1G
77565-900EA 95 USD
77565-900
1-Bromo-4-(trimethylsilyl)but-3-yn-2-one 95%
1-Bromo-4-(trimethylsilyl)but-3-yn-2-one

Precaución: For research use only.

Formula: C₇H₁₁BrOSi
MW: 219.1511 g/mol
MDL Number: MFCD24464079
CAS Number: 433257-41-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR