Methyltransferase Assay Buffer II

Supplier: Sino Biological

M02-09-01 M02-09-05 M02-09-20
77559-348EA 17 USD
77559-348 77559-350 77559-352
Methyltransferase Assay Buffer II
Buffers Assay Buffers

Stock solution of the buffer used for assaying protein methyltransferases.


Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

Order Now

SPECIFICATIONS

Environmentally Preferable
Formulation Frozen solution of buffer containing: 250 mM Tris-HCl, pH 8.0 and 50 ng/µl BSA
Storage Temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR