Kinase Assay Buffers III

Supplier: Sino Biological

K03-09-01 K03-09-05 K03-09-20
77558-734EA 17 USD
77558-734 77558-736 77558-738
Kinase Assay Buffers III
Buffers Assay Buffers

The 5X Kinase assay buffer III is used for assaying kinase protein.


For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Formulation Frozen solution of buffer containing: 200 mM Tris-HCl, pH 7.4, 100 mM MgCl₂ and 0.5 mg/ml BSA. Add to final concentration of 0.25 mM DTT to the aliquot prior to use. Supplied in separate vial.
Form Liquid
Storage Temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR