Fmoc-Lys(Me)₂-OH·HCl 98%

Supplier: Ambeed
Warning

Synonyms: N-α-Fmoc-N-ε-dimethyl-N-L-lysine hydrochloride, Fmoc-Lys(Me2)-OH.HCl

A967219-100MG A967219-1G A967219-250MG A967219-5G
77007-198EA 23 USD
77007-198 77007-200 77007-202 77552-794
Fmoc-Lys(Me)₂-OH·HCl 98%
Fmoc-Lys(ME)2-OH·HCl

Precaución: for Research Use Only

Formula: C₂₃H₂₉ClN₂O₄
MW: 432.945 g/mol
MDL Number: MFCD05662353
CAS Number: 252049-10-8
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR