2-(Pyridin-2-yldisulfanyl)ethanamine ≥95%

Supplier: Ambeed
Warning

A446494-250MG
77551-344EA 242 USD
77551-344
2-(Pyridin-2-yldisulfanyl)ethanamine ≥95%
2-(Pyridin-2-yldisulfanyl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂S₂
MW: 186.297 g/mol
MDL Number: MFCD19706002
CAS Number: 83578-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR