4-(Trifluoromethyl)phenyl isocyanate ≥99%

Supplier: Ambeed
Danger

Synonyms: α,α,α-Trifluoro-p-tolyl isocyanate, 1-Isocyanato-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)phenylisocyanate, p-Trifluoromethylphenylisocyanate

A363400-1G
77551-052EA 14.42 USD
77551-052
4-(Trifluoromethyl)phenyl isocyanate ≥99%
4-(Trifluoromethyl)phenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
MDL Number: MFCD00002028
CAS Number: 1548-13-6
UN: 2285
ADR: 6.1,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR