OneStep™ PLUS qMethyl PCR Kit
Supplier: Zymo Research
D5312
77540-202EA 411.32 USD
77540-202
OneStep™ PLUS qMethyl PCR Kit
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The OneStep PLUS qMethyl PCR Kit provides a simple, straightforward, and bisulfite-free procedure for the quantification of DNA methylation at custom-selected genomic regions.
  • High quality DNA, suitable for use in restriction enzyme digestion
  • A260/280 >1.8, A260/230 >1.8
  • Optimal DNA input is 20 ng in 5 µl
  • Range validated 5 ng to 1 µg (in 5 µl)
Provided dye is compatible with SYBR green filter. Assay is flexible with other dyes and TaqMan probes. Optimal 14 hours of processing time.
DNA in the test reaction is digested with Methylation Sensitive Restriction Enzymes (MSREs) while DNA in the reference reaction is not. The DNA from both reactions is then amplified using Real-Time PCR. The difference in Cycle Threshold (Ct) values between the reference and test reactions is used to calculate the percentage of DNA methylation at the selected genomic region.