ProKlenz® RESTORE High Performance Acid Based Cleaners
Supplier: Steris
142301WR 142305WR
77531-592EA 3338.4 USD
77531-592 77531-590
ProKlenz® RESTORE High Performance Acid Based Cleaners
Cleaning Agents Detergents and Surface Cleansers
ProKlenz® RESTORE Cleaner has an acidic pH for the removal of type I rouge and all-purpose cleaning.
  • Standardization - Globally available product enables harmonized cleaning protocols across sites
  • Sustainability - Free rinsing formula minimizes utility consumption and detergent residue
  • Effectiveness - An acidic pH removes type I rouge while a low foaming system supports use in high-impingement systems
  • Efficiency - A low use-concentration supports cost-effective and optimized cleaning
  • Quality - Manufactured within an FDA registered plant, ISO 9001:2015 and
More Product Information