PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
10TK03 10TK04 10TK05 10TK06 10TK07 10TK08 10TK09 10TK12 10TK16 10TK18
77531-078CS 1815 USD
77531-078 77531-062 77531-068 77531-064 77531-074 77531-070 77531-076 77531-080 77531-066 77531-072
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Covers
PureClean™ Autoclavable Muff Covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® for maximum breathability during sterilization
  • Eliminate the risk of residue deposits on equipment post sterilization
  • Manufactured and packaged in a controlled cleanroom
More Product Information