AccuTec Pro Single Edge Blade Mini-Scraper
Supplier: Accutec Blades
APTL-7002-CJAR
77529-516CS 206.92 USD
77529-516
AccuTec Pro Single Edge Blade Mini-Scraper
Razors and Blades
AccuTec Pro's compact single edge mini scraper is great for quick fixes and scraping.
  • Lightweight and compact
  • Includes one, preinstalled single edge blade
  • Blade can be reversed for safe storage
One mini-scraper per bag, 100 bags per cookie jar.
Precaución: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.