cAMP (Cyclic Adenosine Monophosphate) ELISA Kit

Supplier: AFG BIOSCIENCE LLC

EK247542
77525-500EA 385.7 USD
77525-500
cAMP (Cyclic Adenosine Monophosphate) ELISA Kit
Assays ELISAs

  • Host: General

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein cAMP (Cyclic Adenosine Monophosphate)
Primary antibody reactivity
Assay Type (ELISA with LOV) Competitive
Host
Sensitivity 92.5 pg/ml
Detection Range 312.5 - 20000 pg/ml
Sample Type Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
Time to Results 2 h
Regulatory Status RUO
Storage Temperature 2…8 °C
Shelf Life 6 months (4 °C); 12 months (–20 °C)
Application Signal transduction, Metabolic pathway

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR