Supplier: RPI

P52380-1
77510-412EA 87.4 USD
77510-412
Cell Culture Supplements and Additives

This kit is a special mixture of five protease inhibitors with broad spectrum for the inhibition of various proteases and aminopeptidases. This kit is for use in bacterial cell extracts.


  • AEBSF, HCl: 20 mM, Target: Serine proteases, molecular weight is 239.7
  • Bestatin: 1.7 mM, Target: Aminopeptidase B and Leucin Aminopeptidase, molecular weight is 344.8
  • E-64: 200 μM, Target: Cysteine proteases, molecular weight is 357.4
  • EDTA Disodium: 85 mM, Target: Metalloproteases, molecular weight is 372.2
  • Pepstatin A: 2 mM, Target: Aspartic Proteases, molecular weight is 685.9
  • BSE/TSE free


Appearance: This product is provided as a lyophilized solid, ready for reconstitution. It is provided as a set: one vial with the solid inhibitors (green cap), one 5 ml vial with EDTA and one vial with 1 ml DMSO (colorless cap).


Preparation: Dissolve the solid inhibitors with 1 ml DMSO. Dilute the EDTA vial with 4 ml of distilled water. Add the 1 ml solution of the inhibitors to the EDTA. Mix gently, the solution is cloudy. Optimum titration to be determined in user's own system.


Precaución: Research or further manufacturing use only, not for food or drug use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR