Erlenmeyer Flasks, PETG, Flat Bottom
Supplier: NEST SCIENTIFIC
781001 781011 783001 783011 784001 784011 782001 782011
77508-804CS 111.72 USD
77508-804 77508-806 77508-836 77508-838 77508-852 77508-854 77508-820 77508-822
Erlenmeyer Flasks, PETG, Flat Bottom
Flasks Erlenmeyer Flasks
Ideal for shaker culture applications. PETG Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
  • PETG Bottle and HDPE cap
  • Sterilized by E-Beam
  • Nonpyrogenic
  • Packaged in sterile bags
Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination.
Certificaciones: ISO 9001 and 13485 certified.