Petri Dishes, Sterile
Supplier: NEST SCIENTIFIC
715011 722011 706011 753031 754001 755001 752401 753001 753011 753021
77508-604CS 190.11 USD
77508-604 77508-628 77508-552 77508-748 77508-750 77508-752 77508-740 77508-742 77508-744 77508-746
Petri Dishes, Sterile
Culture Dishes Petri Dishes
Petri dishes are shallow transparent dishes that can be used to grow single cellular organisms, most popularly bacteria.
  • Made of high clarity, 100% virgin polystyrene
  • Sterilized by E-beam, SAL=10⁻⁶
  • Flat, transparent surface
  • Stackable for easy storage and handling
  • Non-pyrogenic, DNase/Rnase free
They are perhaps one of the most commonly used supplies in any laboratory setting. Nest Scientific petri dishes come pre-sterilized, are made of high clarity polystyrene and come with lids. They have good heat resistance and are suitable for use with hot agar.More Product Information