Microcentrifuge Tubes, Polypropylene
Supplier: NEST SCIENTIFIC
615001 615401 615601 620011 620611 603011 603111 605001 605401 605501 605601
77508-356CS 201.16 USD
77508-356 77508-358 77508-360 77508-414 77508-416 77508-234 77508-236 77508-238 77508-240 77508-242 77508-244
Microcentrifuge Tubes, Polypropylene
Tubes Centrifuge Tubes
Microcentrifuge tubes are commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields.
  • Non-pyrogenic
  • DNase/RNase free and deemed animal free
  • Resistant to chemical corrosion and low temperature
  • Clear graduations and frosted marking area
  • Withstand temperature ranging from –80 to 121 ℃
Available in two colors: Amber and clear. Amber centrifuge tubes can also be used to store photosensitive samples and special chemiluminescence reagents.
Ultra-high centrifugal stability, centrifugalMore Product Information