Cell Culture Chamber Slides
Supplier: NEST SCIENTIFIC
230101 230102 230104 230108 230111 230112 230114 230118 230121 230122 230124 230128
77507-434CS 108.32 USD
77507-434 77507-436 77507-438 77507-440 77507-442 77507-444 77507-446 77507-448 77507-450 77507-452 77507-454 77507-456
Cell Culture Chamber Slides
Slides Chamber Slides
NEST Cell culture chamber slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocyto-logical experiments.
  • Convenient for microscopic observation
  • Bottom materials: Borosilicate glass, thickness 1.0 to 0.1 mm, no surface treatment for glass bottom
  • Chamber color: Clear/white/black
  • Easy open flip for chamber and slide disassembly
  • No chemical adhesives used
  • Designed to prevent cross contamination, sterile
  • Alphanumeric labeling
Cell culture chamber slides provide removable chambers that enable cellMore Product Information