Formalin, VWRClick, neutral buffered prefill

Supplier: VWR International
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

77507-050 77507-052 77507-054 77507-056 77507-058 77507-042 77507-044 77507-046 77507-048 77507-040 77507-060
77507-050CS 184.63 USD
77507-050 77507-052 77507-054 77507-056 77507-058 77507-042 77507-044 77507-046 77507-048 77507-040 77507-060
Formalin, VWRClick, neutral buffered prefill
Formalin

VWR®, part of Avantor has industry leading Anatomical Pathology prefills available for your lab.


  • VWRClick containers produce an audible “click” sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.
  • Packaged in HistoPack carrier for easy distribution or in bulk.
  • Containers are 95kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.
  • "Click" available in 20 ml, 40 ml, 60 ml, 90 ml, 120 ml sizes.


Certificaciones: ISO 13485 Certified

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Melting Pt: –92 °C (–133 °F)
Flash Pt: 93.3 °C (199.9 °F)
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR