Automatic Liquid Handling Syringes, XLP
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2679611 2679613 2679711 2679713 2679811 2679813 2679911 2679913 2679091 2679095 2679101
77506-548EA 82.42 USD
77506-548 77506-550 77506-552 77506-554 77506-556 77506-558 77506-560 77506-562 77506-542 77506-544 77506-546
Automatic Liquid Handling Syringes, XLP
Syringes Syringes for Chromatography
Series XLP Automatic Liquid Handling syringes with a glass barrel made from Borosilicate glass 3.3 designed for volumes of 25 µl to 50 ml.
  • Luer or ¼-28 threaded end caps
  • Broad variety of syringe interfaces
  • Seals available in PTFE, UHMWPE, or custom UHMWPE blends
  • Customized designs and materials to suit application requirements
  • Stroke 60 mm
A high chemical resistance makes this syringe a perfect heavy-duty syringes. The syringe is heat and shock resistant, which ultimately protects against breakage from shock and sudden or extreme fluctuations inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable