4-(4-Bromophenyl)tetrahydropyran ≥97%

Supplier: AOBChem USA

Warning

26861-1G 26861-500MG 26861-5G 26861-25G
77467-862EA 116.39 USD
77467-862 77480-562 77490-902 77500-962
4-(4-Bromophenyl)tetrahydropyran ≥97%
4-(4-Bromophenyl)tetrahydropyran

Precaución: For research use only

Formula: C₁₁H₁₃BrO
MW: 241.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913643
CAS Number: 811828-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR