2-Fluoro-6-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-6-methoxybenzeneboronic acid

10469-25G
77497-526EA 64.38 USD
77497-526
2-Fluoro-6-methoxyphenylboronic acid ≥97%
2-Fluoro-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179483
CAS Number: 78495-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR